missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Dynabeads™ MAX E. coli O157 Kit
Dynabeads™ MAX E. coli O157 Kit
POA
Specifications
For Use With (Equipment) | Dynabeads™ MPC™, KingFisher mL Food Protection Purification System, DynaMag™-2 |
---|---|
Product Type | E.coli Kit |
Sample Type | Environmental Sample, Feces, Food, Water Samples |
Description
Dynabeads™ MAX E.Coli O157 provides a powerful sample preparation method to improve PCR detection and quantification of E.Coli O157:H7 in food, feces, water and other environmental samples.Based on well known and widely used immunomagnetic separation (IMS) technology, Dynabeads™ MAX E.Coli O157 provides a selective E.Coli O157:H7 capture step that delivers significant improvements in PCR analytical process time, as well as the sensitivity and consistency of results. With Dynabeads™ MAX E.Coli O157, you'll:
Specifications
Dynabeads™ MPC™, KingFisher mL Food Protection Purification System, DynaMag™-2 | |
E.coli Kit | |
Environmental Sample, Feces, Food, Water Samples |
For Research Use Only. Not for use in diagnostic procedures.