missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Dynabeads™ anti-E. coli O157
Dynabeads Anti-E. coli O157
£326.00 - £804.00
Specifications
Product Type | Anti-E.coli O157 Bead |
---|---|
For Use With (Equipment) | Dynabeads™ MPC™, KingFisher mL Food Protection Purification System, DynaMag™-2 |
Isolation Technology | Immunomagnetic Separation |
Prep Scale | Small Volume Microbial Samples |
Product Line | Dynabeads™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10322443
|
Applied Biosystems™
71003 |
1 mL |
£326.00
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
11593367
|
Applied Biosystems™
71004 |
5 mL |
£804.00
5mL |
In Stock
Log in to see stock. |
|||||
Description
Dynabeads™ E. coli anti-O157 is a tool for better sample preparation and improved cell culture and immuno-based assays for E. coli O157:H7 in food, feces, water and other environmental samples. Based on well-known and widely used immunomagnetic separation (IMS) technology, Dynabeads™ E. coli anti-O157 provides a useful means to selectively capture E. coli O157:H7 and better prepare samples for effective analysis.With Dynabeads™ anti-E. coli O157, you will:
Specifications
Anti-E.coli O157 Bead | |
Immunomagnetic Separation | |
Dynabeads™ |
Dynabeads™ MPC™, KingFisher mL Food Protection Purification System, DynaMag™-2 | |
Small Volume Microbial Samples | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.