Learn More
Thermo Scientific™ DyLight™ Yellow Specialty Dyes
Label amine-containing molecules for fluorescence imaging or flow cytometry with these rhodamine-based, yellow-emitting (532 to 546 laser) NHS ester fluorescent dyes.
Brand:  Thermo Scientific™ 46614
Code : PN
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12341903
  • £222.07 / 1mg
Estimated Shipment
10-07-2018
Add to basket
Description
Description
DyLight Yellow-emitting Dyes are a family of labeling agents that provide bright fluorescent detection for imaging. Dyes can be selected based upon their characteristic excitation and emission properties or relative hydrophilicity & hydrophobicity attributes. Dyes that contain a greater number of negatively charged sulfonates generally will have greater water solubility than dyes with fewer sulfonates. More hydrophobic dyes often provide better cell penetrating ability in vivo, while more hydrophilic dyes have less nonspecific binding potential. Each dye contains an amine-reactive NHS ester for simple modification of antibodies, proteins, peptides or other biomolecules through amide bond formation.
Highlights:
  • NHS ester reactive group–allows immediate labeling of antibodies, proteins, peptides and other amine-containing molecules through amide bond formation
  • Multiple solubility options –choose from hydrophilic to hydrophobic dyes to optimize the right dye label for the best performance in a given application
Criteria to consider when choosing a DyLight Yellow-emitting Specialty Dye:
  • Excitation and emission wavelengths–choose the best dye to match the excitation and emission capabilities of your instrument
  • Water solubility–choose a dye based on its relative hydrophilicity, which directly correlates to the number of negatively-charged sulfonates it has on its core structure. More hydrophilic dyes are best at maintaining water solubility of a labeled antibody and limiting the nonspecific binding of the conjugate. More hydrophobic dyes often are best at penetrating tissues and cell membranes in vivo, meaning that dyes with fewer sulfonates may work best for some applications.
Recommended for:
Fluorescence imaging; Fluorescence correlation spectroscopy; Flow cytometry; Imaging; Antibody labeling; Direct immunofluorescence staining; ELISA; Western blotting; Protein microarrays; Polymer labeling; Peptide labeling; Phalloidin labeling for actin staining
Specifications
Specifications
DyLight 554-R1 NHS Ester
NHS ester activated Rhodamine derivative with 1 sulfonate group
1 use to label a total of 7mg of IgG using typical conditions