Learn More
Thermo Scientific™ DyLight™ Long Stokes Shift Specialty Dyes
Label antibodies or other proteins for FRET and multiplex fluorescence assays using fluors with large Stokes shifts (488nm excitation; yellow to far-red emission).
£237.00
Specifications
Description | DyLight 485-LS NHS Ester |
---|---|
Formulation | Long stokes shift NHS ester activated Coumarin derivative |
Quantity | 1 mg |
Sufficient For | 1 use to label a total of 7mg of IgG using typical conditions |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
12351943
|
Thermo Scientific™
82492 |
DyLight 485-LS NHS Ester |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Thermo Scientific™ DyLight Long Stokes Shift Specialty Dyes offer a broad spectrum of fluorescence emission characteristics from green to far red with a single excitation peak that matches the 488 laser line and extraordinarily large Stokes shifts that inhibit quenching.
DyLight Long Stokes Shift Specialty Dyes are a family of labeling agents that provide bright fluorescent detection for multiplexed imaging. The dyes share a common excitation range, while their emission properties extend from the green to the far red wavelengths. The dyes are based on a core coumarin structure that is modified to emit at different wavelengths. Each dye contains an amine-reactive NHS ester for rapid modification of antibodies, proteins, peptides or other biomolecules through amide bond formation. DyLight Long Stokes Shift Dyes can be used individually or combined with DyLight 488 for multiplexed analysis. The excitation and emission peaks are sufficiently separated so as to minimize dye-dye quenching in assays.Highlights:
Single excitation wavelength–DyLight Long Stokes Shift Dyes all can be excited together at 488nm using an Argon laser
Image multiple targets simultaneously–the four Long Stokes Shift Dyes can be used with DyLight 488 for multiplex fluorescence applications
NHS ester reactive group–allows immediate labeling of antibodies, proteins, peptides and other amine-containing molecules through amide bond formation
Multiple solubility options –choose from hydrophilic to hydrophobic dyes to optimize the right dye label for the best performance in an application
Large Stokes shift inhibits quenching–minimal overlap between excitation and emission peaks inhibits dye-dye quenching effects
Use in FRET-based assays– the extremely broad separation between excitation and emission peaks allow for use of the Long Stokes Shift Dyes in energy transfer applications where the acceptor dye is spectrally far removed from the excitation wavelength
Criteria to consider when choosing a DyLight Long Stokes Shift Specialty Dye:
- Excitation and emission wavelengths–all dyes share a common excitation wavelength range with options for emission from the green to the far red wavelengths
- Water solubility–DyLight Long Stokes Shift Dyes can be initially solubilized in methanol, ethanol, DMF or DMSO with transfer into an aqueous buffer medium for labeling biomolecules
Recommended for:
Quenchers and FRET pairs; Conjugation to silica particles; STED microscopy; Multicolor microscopy; 4Pi microscopy; Dual-color 3D nanoscopy; Dynamic light scattering; Graded stokes shift; Duplex enhanced RT-PCR; High-resolution optical microscopy
Specifications
DyLight 485-LS NHS Ester | |
Long stokes shift NHS ester activated Coumarin derivative | |
1 mg | |
1 use to label a total of 7mg of IgG using typical conditions |