Learn More
Thermo Scientific™ DyLight™ In Vivo Apoptosis Detection Kits and Reagents
Measure drug-induced changes in apoptosis in vivo (live animal imaging) using near-infrared DyLight Dye fluorescent polycaspase probes.
Brand:  Thermo Scientific™ 84889
Code : PN
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12311943
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
These kits contain a DyLight 690- or DyLight 747-conjugated in vivo polycaspase probe, which is a cell-permeable fluorescent detector of active caspases indicative of apoptosis. The probes are comprised of a polycaspase peptide sequence (VAD) and a fluoromethyl ketone (FMK) moiety, which facilitate irreversible binding of the probe to active caspases, and either DyLight 690 or DyLight 747 Dye for near-infrared detection with a range of in vivo instrumentation. These probes are valuable for in vivo drug determination studies, because changes in apoptosis from drug reactivity can be measured by fluorescence imaging in live animals. An increase in apoptosis produces a greater attachment of probes and, subsequently, an increase in fluorescence output. The in vivo technique is nondestructive, thus eliminating the need to remove cells and tissues from sacrificed animals prior to analysis (which induces further apoptosis). Additionally, apoptosis can be monitored in a single animal at multiple time points, reducing the number of animals required for a given study.
Highlights:
  • Nontoxic–not harmful to valuable research animals
  • Cell-permeable–readily diffuses through all cells as it circulates throughout the body
  • Selective for active caspases–eliminates high background and false-positive results common with other caspase detection methods
Recommended for:
Small-animal in vivo apoptosis imaging
Specifications
Specifications
DyLight 690 In Vivo Control Reagent
Proprietary dark green lyophilized powder
10 tests per vial
1 vial
Videos