Learn More
Thermo Scientific™ DyLight™ 680-4xPEG Amine-Reactive Dye and Kits
Fluorescently label antibodies and other proteins with Thermo Scientific™ DyLight 680-4xPEG Amine-Reactive Dye, a derivative of our high-performance DyLight 680 Dye.
Brand:  Thermo Scientific™ 53076
Code : PN
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13444219
  • £324.00 / Each
Estimated Shipment date
from Supplier 29-10-2019
Description
Description
The DyLight 680-4xPEG dye contains 4 polyethylene glycol (PEG) chains that are non–cytotoxic, enhance fluorescence and reduce nonspecific binding of conjugates made with them. Conjugates made with DyLight 680-4xPEG Dye can be used as molecular probes for cellular imaging and other fluorescence detection methods. The far-red to NIR fluorescence properties of DyLight 680-4xPEG Dye make it especially useful in a variety of biological, chemical, and pharmaceutical applications, including in vivo imaging. The PEG chains also improve solubility of the dyes and labeled molecules in aqueous solution, aid in cell permeability and improve tissue retention.
Highlights:
  • High fluorescence intensity – significantly brighter fluorescence than Alexa Fluor™ 680
  • PEGylated – improves solubility in aqueous solution, aids in cell permeability and improves tissue retention in in vivo applications
  • Convenient kit sizes – standard and microscale kit sizes to match your experimental needs
Recommended for:
  • Fluorescence microscopy
  • In vivo or ex vivo imaging
  • Cell-based assays
  • Flow cytometry/fluorescence-activated cell sorting (FACS)
Specifications
Specifications
DyLight 680-4xPEG Antibody Labeling Kit
Borate Buffer (0.67M), 1mL, DyLight Fluor NHS Ester, 3 vials, Microcentrifuge Collection Tubes, 12 tubes, Microcentrifuge Spin Columns, 6 columns, Purification Resin, 5mL
3-rxn kit
Multi-component kit with proprietary near-infrared fluorescent dye
Labeling and purifying 3 x 1mg (2mg/mL) of IgG or other proteins