Learn More
DY131, Tocris Bioscience™
Agonist selective for estrogen-related receptors ERRβ and ERRγ
Brand:  Tocris (Bio-Techne) 2266/10
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13460894
  • £132.30 / 10mg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
Novel selective agonist at estrogen-related receptors ERRβ and ERRγ. Displays minimal activity at ERRα, ERα and ERβ at concentrations up to 30 μM.
Specifications
Chemical Identifiers
95167-41-2
311.385
unii-5vwv92039e, n-4-diethylaminobenzylidenyl-n'-4-hydroxybenzoyl-hydrazine, d06jpq, n'-4-diethylamino phenyl methylidene-4-hydroxybenzohydrazide, n-e-4-diethylamino phenyl methylideneamino-4-hydroxybenzamide, benzoic acid, p-hydroxy-, p-diethylamino benzylidene hydrazide, n'-1e-4-diethylamino phenyl methylene-4-hydroxybenzohydrazide, benzoic acid, 4-hydroxy-, 4-diethylamino phenyl methylene hydrazide, benzoic acid, 4-hydroxy-, 2-4-diethylamino phenyl methylene hydrazide, dy hplc
N-[(E)-[4-(diethylamino)phenyl]methylideneamino]-4-hydroxybenzamide
C18H21N3O2
WLKOCYWYAWBGKY-CPNJWEJPSA-N
5497124
CCN(CC)C1=CC=C(C=C1)C=NNC(=O)C2=CC=C(C=C2)O
Specifications
DY131
95167-41-2
unii-5vwv92039e, n-4-diethylaminobenzylidenyl-n'-4-hydroxybenzoyl-hydrazine, d06jpq, n'-4-diethylamino phenyl methylidene-4-hydroxybenzohydrazide, n-e-4-diethylamino phenyl methylideneamino-4-hydroxybenzamide, benzoic acid, p-hydroxy-, p-diethylamino benzylidene hydrazide, n'-1e-4-diethylamino phenyl methylene-4-hydroxybenzohydrazide, benzoic acid, 4-hydroxy-, 4-diethylamino phenyl methylene hydrazide, benzoic acid, 4-hydroxy-, 2-4-diethylamino phenyl methylene hydrazide, dy hplc
CCN(CC)C1=CC=C(C=C1)C=NNC(=O)C2=CC=C(C=C2)O
311.385
311.38
10mg
C18H21N3O2
WLKOCYWYAWBGKY-CPNJWEJPSA-N
N-[(E)-[4-(diethylamino)phenyl]methylideneamino]-4-hydroxybenzamide
5497124
>99%
Videos