Learn More
Ansell™ Duzmor™ Plus 87-600 Series Natural Rubber Latex Immersion Gloves
A unique combination of strength and elasticity for safer handling of more hazardous substances. Only 0.43 mm thick for outstanding sensitivity and tactile performance.Reduced risk of allergic reactions.
Brand: Ansell™ 87600090BP
17.36 GBP valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.77400kg
Description
- Made from unsupported natural rubber
- Reduces risk of allergic reactions
- Unflocked liner
- Fishscale grip
- Bisque fingertip grip
- Pinked cuff
Specifications
Rubber Latex | |
Natural Rubber Latex | |
Immersion Gloves | |
87-600 | |
Natural | |
Unflocked | |
Hazardous substances |
Natural Rubber Latex | |
No | |
9 | |
0.43 mm | |
Pinked | |
12 Pairs | |
87-600 |