Learn More
DWK Life Sciences DUROPLAN™ Petri Dish, lid and base have planar surfaces, and free from bubbles and streaks
Petri dishes are used for the cultivation of bacteria, yeasts, plant and animal cells on solid media.
Brand: DWK Life Sciences 217555303
262.98 GBP valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Code : A0
Additional Details : Weight : 2.16900kg
Description
Cultivation of microorganisms can take place on a gel-like nutrient medium, which is placed in a flat, round, transparent dish with lid (the so-called Petri dish). Due to the solid gel, mixing of the microorganisms is prevented and larger cell culture accumulations (colonies) can grow. DURAN™ Petri dishes are manufactured from borosilicate glass 3.3 using a special manufacturing process. They allow an even distribution of the culture medium and guarantee a distortion-free view. The lids of the DURAN™ Petri dishes rest directly on the lower part and thus prevent turbulence of the protruding air space. This is well suited for reactions with long incubation times, at high temperatures or with sensitive culture media.
- Special manufacturing process permits the uniform distribution of agar and guarantees distortion-free observation
- Bottom and lid are microscopically flat on the outside and inside
- Free from bubbles and streaks
Specifications
Autoclavable | |
Growth and cultivation of microorganisms; Cultivation on solid gel culture medium; Microscopy of microorganisms; Cell culture | |
32 mm |
Borosilicate Glass (3.3) | |
10/Pk. | |
150 mm |