missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ TILT Bottle Tag, GL 56, Silicone Rubber

Product Code. p-7189105 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Orange
Purple
Yellow
Unit Size:
Pack of 20
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15503547 Orange Pack of 20
15513547 Yellow Pack of 20
15523547 Blue Pack of 20
15533547 Purple Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15503547 Supplier DWK Life Sciences Supplier No. 292435626
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Simple colour coding for DURAN™ TILT bottles.

The Bottle Tags can be used on their own for colour identification or to secure the protective Light Shield around the bottle. The GL 56 Bottle Tags are available in four colours: orange, yellow, blue and purple.

  • The Bottle Tags can be used for colour identification or to secure the protective Light Shield around the bottle
  • The elastic Bottle Tags can be easily and securely attached around the neck, even with the screw cap in place
  • Feature a graspable tab that makes them easy to remove with bare or gloved hands
  • Chemically robust, heat resistant and reusable

Specifications

Color Orange
Material Silicone
For Use With (Application) color coding and identification; Attachment of the protective Light Shield
No. per Pack 20/Pk.
Thread Size GL 56
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.