Learn More
DWK Life Sciences DURAN™ Standard Socket, diamond-ground
For a rigid, self-supporting connection as a cost-effective alternative.
Brand: DWK Life Sciences 286532806
Additional Details : Weight : 1.25000kg
Description
The conical ground joint of sleeve and core is a rigid, self-supporting joint. The DURAN™ DG sleeves are not ground according to DIN 12249 and represent a cost-effective alternative to the DURAN™ sleeves. In principle, ground joints should be made using ground grease or PTFE sleeves. KECK™ Conical ground joint clamps are the perfect supplement for securing the conical ground joint.
- Diamond cut (coarser cut quality, more roughness depth)
- Cost-effective alternative to DURAN™ sleeves (not according to DIN)
- Tight closure due to standard ground joint
- Glass articles for further processing
Specifications
50/Pk. | |
Use in the high vacuum range; Installation in glass apparatus and devices | |
17 mm | |
1.8 mm |