Learn More
DWK Life Sciences DURAN™ Receiver Adapter, bent, with standard ground cone
The receiver adapter is suitable for transferring the distillate from the condenser to the receiving flask.
Brand:  DWK Life Sciences 243100808
Code : NEW
Additional Details : Weight : 0.03200kg
 View more versions of this product
Product Code. 10131791
  • £18.44 / Each
Estimated Shipment: 22-11-2021
Add to basket
Description
Description
Part of a distillation assembly is, among other things, the receiver adapter, which is connected downstream at the end of the condenser and guides the distillate into the receiving flask.
  • Tight closure due to standard ground joint
  • Bent
  • With standard ground socket
  • Fired-on, highly durable white print
Specifications
Specifications
1 Ea.
Distillation
NS 24/29