Learn More
DWK Life Sciences DURAN™ Filter Disc, fused glass rim
The glass filter disk is used to collect or separate solids from the solution to be filtered.
£30.05 - £102.00
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | Qualitative inorganic analysis and preparative chemistry; Filtration without filter paper; Filtering |
No. per Pack | 1/Pk. |
Product Code | Brand | Diameter (Metric)Top | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Description | Price | Quantity & Availability | ||||
10169370
|
DWK Life Sciences
252024104 |
24 mm Plate | Porosity 1 |
£30.05
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10797931
|
DWK Life Sciences
252024207 |
24 mm Plate | Porosity 2 |
£30.05
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10078090
|
DWK Life Sciences
252024301 |
24 mm Plate | Porosity 3 |
£30.05
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
10528291
|
DWK Life Sciences
252024404 |
24 mm Plate | Porosity 4 |
£32.45
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
10528481
|
DWK Life Sciences
252050105 |
50 mm plate | Porosity 1 |
£42.25
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
10473131
|
DWK Life Sciences
252050208 |
50 mm plate | Porosity 2 |
£50.35
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
||||
10614671
|
DWK Life Sciences
252050302 |
50 mm plate | Porosity 3 |
£42.25
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
10119520
|
DWK Life Sciences
252050405 |
50 mm plate | Porosity 4 |
£45.05
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||||
10179560
|
DWK Life Sciences
252090109 |
90 mm plate | Porosity 1 |
£102.00
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||||
10149660
|
DWK Life Sciences
252090203 |
90 mm plate | Porosity 2 |
£67.95
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||||
10729281
|
DWK Life Sciences
252090306 |
90 mm plate | Porosity 3 |
£56.70
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
10716981
|
DWK Life Sciences
252090409 |
90 mm plate | Porosity 4 |
£60.95
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
Description
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. The filter funnel head consisting of the filter head, sealing ring, glass filter disk and funnel can be dismantled into its individual parts and each individual component can be replaced or varied in order to adapt the filter to the application. The three unit sizes combined with four filter plates of different porosities each create 12 different filter options.
- If a glass filter is used, a filter paper or membrane may not be required
- With fused glass rim
- Made of DURAN™ glass, with its proven characteristics, such as chemical resistance and resistance to thermal shock.
Specifications
Borosilicate Glass (3.3) | |
Qualitative inorganic analysis and preparative chemistry; Filtration without filter paper; Filtering | |
1/Pk. |