Learn More
DWK Life Sciences DURAN™ Filter Apparatus, Witt Type, complete, with interchangeable lid, and KECK™ assembly set, standard ground joint 29/32
The Witt filtration apparatus is used for solid-liquid filtration in which the filtrate is subsequently further processed.
Brand: DWK Life Sciences 247304603
299.39 GBP valid until 2024-12-31
Use promo code "21897" to get your promotional price.
Code : NEW
Additional Details : Weight : 1.15000kg
Description
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Buechner funnels are cylinders with an integrated large-pored sieve plate. Without additional filter paper, only coarse filtration is possible. Buechner funnels can also be used in vacuum filtration. The applied vacuum accelerates the filtration.
- Tight closure due to standard ground joint
- With interchangeable lid and KECK™ assembly set
- Fired-on, highly durable white print
Specifications
Autoclavable | |
Borosilicate Glass (3.3), PBT, Polypropylene, VMQ, EPDM | |
1 Each |
NS 29/ 32, DN 100 | |
Filtering; Preferably collecting the filtrate; Filtration at reduced pressure; Vacuum filtration; Safe working under vacuum |