missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Duplex 2209, 40 mesh woven from 0.2mm (0.008mm) dia wire

Brand:  Alfa Aesar™ 046504.GT

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 150 x 150mm

 View more versions of this product

Product Code. 15407235

  • £54.40 / Each


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

Duplex 2209
40 mesh woven from 0.2mm (0.008mm) dia. wire

Specifications

Duplex 2209
150 x 150mm
Odorless
40 Mesh
40 mesh woven from 0.2mm (0.008mm) dia. wire
Wire Cloth
Videos
SDS