missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dual-Light™ Luciferase & β-Galactosidase Reporter Gene Assay System
Rapid and sensitive detection of firefly luciferase and E. coli β-galactosidase in a single extract aliquot for gene expression assays.
£357.00 - £3400.00
Specifications
Product Type | β-Galactosidase/Luciferase Reporter Gene Assay System |
---|---|
Detection Method | Chemiluminescent |
For Use With (Equipment) | Luminometer (Microplate), Luminometer (Tube-Based) |
Assay | β-Gal (lacZ) Assay, Luciferase Assay |
Content And Storage | This kit contains the enzyme substrate, lysis solution, required buffers, and reaction acccelerator. Store the product at -20°C. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10055924
|
Invitrogen™
T1003 |
200 Assays |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12654265
|
Invitrogen™
T1004 |
4000 Assays |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12664265
|
Invitrogen™
T1005 |
600 Assays |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Detection of firefly luciferase and β-galactosidase in same sample simplifies normalization of transfection efficiency
- Wide dynamic range of β-galactosidase and luciferase assays enables accurate measurement of both enzymes from femtogram to nanogram range
- Assay sensitivity is 100- to 1,000-fold better than either the isotopic/non-isotopic assays for chloramphenicol acetyl transferase (CAT) or the colorimetric/fluorescent assays for β-galactosidase, providing greater sensitivity than competing assay technologies
- Highly sensitive assay with a wide dynamic range permits detection of high and low levels of reporter without performing numerous sample dilutions
- Non-radioactive reporter gene assay kit eliminates concerns over use of radioisotopes
- Assay can be completed in about one hour, providing fast assay turnaround
Easy-to-Use
Light signal from each enzymatic reaction is measured sequentially in a luminometer with automatic injectors or other instrumentation in which light emission measurements can be performed within a short period. First, luciferase reporter enzyme activity is quantitated with an enhanced luciferase reaction. Following a 30-60 minute incubation and addition of a light-emission accelerator, β-galactosidase reporter enzyme activity is determined with Galacton-Plus™ substrate. Both assays are combined into one simple assay using only one extract aliquot for greater convenience and precision. The entire assay is completed in less than one hour.
Wide Dynamic Range
The wide dynamic range of this dual assay enables accurate measurement of luciferase and β-galactosidase concentrations over seven orders of magnitude, from the femtogram to nanogram range. Colorimetric and isotopic reporter gene assays cannot rival the dynamic range of the Dual-Light system. We have formatted this assay for tube or microplate luminometer, with injection capability.
Specifications
- Compatible Cells: Mammalian Cells, Yeast Cells
- Detection Method: Chemiluminescent
- For Use With Equipment: Luminometer (Microplate), Luminometer (Tube-Based)
- Form: Liquid
- Format: 96-well plate
- High Throughput Compatibility: High Throughput-Compatible
- Readout: End Point
- Sample Type (Specific): Cell Cultures
- Substrate: Galacton-Plus, Luciferin
- Substrate Properties: Chemical Substrate
- Substrate Type: Beta-Gal Substrate, Luciferase Substrate
Specifications
β-Galactosidase/Luciferase Reporter Gene Assay System | |
Luminometer (Microplate), Luminometer (Tube-Based) | |
This kit contains the enzyme substrate, lysis solution, required buffers, and reaction acccelerator. Store the product at -20°C. |
|
High-throughput Compatible | |
Cell Cultures | |
Beta-Gal Substrate, Luciferase Substrate | |
Luciferase, Beta-Galactosidase | |
Enzyme Assay |
Chemiluminescent | |
β-Gal (lacZ) Assay, Luciferase Assay | |
Mammalian Cells, Yeast Cells | |
End Point | |
Galacton-Plus™, Luciferin | |
Chemical Substrate | |
Dual-Light™, NovaBright™ | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.