missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dtk Rat anti-Mouse, Clone: 109646, R&D Systems™
Rat Monoclonal Antibody has been used in 6 publications
POA
Specifications
Antigen | Dtk/TYRO3 |
---|---|
Clone | 109646 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Tyro3/Dtk Ala31-Ser418 Accession # P55144 |
Gene ID (Entrez) | 7301 |
Gene Accession No. | P55144 |
Description
Dtk/TYRO3 Monoclonal specifically detects Dtk/TYRO3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
Dtk/TYRO3 | |
Mouse myeloma cell line NS0-derived recombinant mouse Tyro3/Dtk Ala31-Ser418 Accession # P55144 | |
P55144 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Rat | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
109646 | |
7301 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects mouse Tyro3/Dtk extracellular domain in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Tyro3/Dtk, recombinant mouse (rm) Mer, or rmAxl is observed. | |
Unconjugated | |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF, ELISA | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only