missing translation for 'onlineSavingsMsg'
Learn More

Genlab Drying/Warming Cabinet w/Fan

Energy-efficient, safe, less expensive glassware drying solution.

£1465.00 - £5345.00

Specifications

Electrical Requirements 220-240 V, 50 Hz, 1 ph.
View More Specs

Products 4
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15658665
View Documents Promotion Details
Genlab
E3DWC100/F
100 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15678665
View Documents Promotion Details
Genlab
E3DWC200/F
200 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15698665
View Documents Promotion Details
Genlab
E3DWC425/F
425 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15618675
View Documents Promotion Details
Genlab
E3DWC885/F
885 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • Timer to automate shut-down periods.
  • Fully insulated with adjustable vent cover.
  • Integral 7-day timer with display.
  • Digital controller with fixed over-temperature safety cut-out.
  • Adjustable shelf runners with removable chrome shelves.
  • Easy to clean, powder-coated body with toughened glass doors.
  • Low energy consumption: ≤50% lower than traditional cabinets.
  • Low heat output: reduces air conditioning costs.
  • Heat up time ≤1 hour.
  • Lockable casters on 425 L and 885 L models.
Specifications

Specifications

220-240 V, 50 Hz, 1 ph.
Videos
SDS
Documents

Documents

Product Certifications