missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ DreamTaq™ Hot Start Green DNA Polymerase
Thermo Scientific DreamTaq Hot Start DNA Polymerase is the product of choice when you need consistently robust amplification with minimal optimization steps
£71.00 - £2360.00
Specifications
GC-Rich PCR Performance | Low |
---|---|
Polymerase | DreamTaq DNA Polymerase |
Reaction Speed | Standard |
Product Line | DreamTaq™ |
Shipping Condition | Wet or Dry Ice |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15629374
|
DreamTaq
EP1711 |
200 U |
£91.00
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15639374
|
DreamTaq
EP1712 |
500 U |
£196.00
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15649374
|
DreamTaq
EP1713 |
2500 U |
£682.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15659374
|
DreamTaq
EP1714 |
4 x 2500 U |
£2360.00
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15646154
|
DreamTaq
EP1711 |
200 U |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15707506
|
DreamTaq
EP1712 |
500 U |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15666154
|
DreamTaq
EP1713 |
2500 U |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15717506
|
DreamTaq
EP1713 |
2500 U |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15605014
|
DreamTaq
EP1713 |
2500 U |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15676154
|
DreamTaq
EP1714 |
4 x 2500 U |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15656154
|
DreamTaq
EP1712 |
500 U |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Thermo Scientific™ DreamTaq™ Hot Start DNA Polymerase is an enhanced hot start Taq DNA polymerase that ensures higher PCR specificity, sensitivity and yields compared to conventional hot start Taq DNA polymerases. DreamTaq Hot Start DNA Polymerase uses antibody based inhibition of DNA polymerase activity at ambient temperatures, thus preventing the amplification of non-specific products prior to the amplification step.Better reaction outcomes:
- Higher yield of target amplicons from low template amounts.
- Increased reaction specificity due to reliable hot-start technology.
- Wider range of amplicon lengths - routine amplification of genomic DNA fragments up to 6kb.
Enhanced convenience:
- Room temperature reaction set-up.
- Minimized optimization of Mg2+ concentration and of primer annealing temperatures due to optimized reaction buffer.
- Direct loading of PCR products on a gel with DreamTaq Green buffer.
Specifications
Low | |
Standard | |
Wet or Dry Ice | |
3'-A |
DreamTaq DNA Polymerase | |
DreamTaq™ | |
Built-In Hot Start | |
Separate Components |