Learn More
Nickel Electro™ Water Bath Drain Tap
Nickel Electro™ Water Bath Drain Tab is designed to fit in all Clifton™ bath drain outlets.
Brand: Nickel Electro™ BX0688
Code : 03
Additional Details : Weight : 1.00000kg
Description
- Permits easy emptying of water baths
- Providing a gated on/off valve to empty fluids conveniently from the bath
- Nozzle supplied with tap for laboratory tubing
- Before fitting make sure bath is empty of liquid. Unscrew the stop plug with a 6mm Allen key whilst holding the nut with a 21mm spanner or nut holder supplied with tap. Screw the Tap into position, recommend pinch tight.
Specifications
Water Baths with Serial Numbers Up to 75100 | |
Drain Tap |