missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DR3/TNFRSF25 Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | DR3/TNFRSF25 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
DR3/TNFRSF25 Polyclonal specifically detects DR3/TNFRSF25 in Human samples. It is validated for Western Blot.Specifications
DR3/TNFRSF25 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
Apo-3, APO3, Apoptosis-inducing receptor AIR, Apoptosis-mediating receptor DR3, Apoptosis-mediating receptor TRAMP, Death receptor 3, DR3member 12 (translocatingchain-association membrane protein), LARD, Lymphocyte-associated receptor of death, Protein WSL, Protein WSL-1, TNFRSF25, TRAMP, tumor necrosis factor receptor superfamily, member 25, WSL, WSL1, WSL-1 | |
TNFRSF25 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q93038 | |
8718 | |
Mouse myeloma cell line NS0-derived recombinant human DR3/TNFRSF25 Gln25-Phe201 Accession # Q93038 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human DR3/TNFRSF25 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse DR3 is observed, and less than 1% cross-reactivity with recombinant human (rh) Fas, rhTNF RI, rhDR6, rh4-1BB, rhCD27, rhCD30, rhCD40, rhGITR, rhHVEM, rhTNF RII and rhOPG is observed. |
For Research Use Only