Learn More
DR3 Mouse anti-Human, PE, Clone: JD3, eBioscience™
Mouse Monoclonal Antibody
£81.50 - £190.00
Specifications
Antigen | DR3 |
---|---|
Clone | JD3 |
Target Species | Human |
Conjugate | PE |
Isotype | IgG1, kappa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15587066
|
Affymetrix eBioscience
12-6603-41 |
25 Tests |
£81.50
25 tests |
Estimated Shipment 03-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15597066
|
Affymetrix eBioscience
12-6603-42 |
100 Tests |
£190.00
100 tests |
Estimated Shipment 03-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The JD3 monoclonal antibody reacts with human DR3, also known as Apo-3, WSL-1, TRAMP, LARD, DDR3, and TR3. DR3, a novel death receptor is expressed at low level by monocytes and granulocytes. Interaction of DR3 with its ligand activates NF-kappa B pathway and induces apoptosis. Initially, DR3 was thought to be a receptor for TWEAK, but further studies have shown that TWEAK could induce apoptosis via receptors distinct from DR3. JD3 has been reported for use in flow cytometric analysis. Applications Reported: The JD3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This JD3 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells and human DR3-transfected cells. This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
DR3/TNFRSF25 is a member of the TNF-receptor superfamily. This receptor is expressed preferentially in the tissues enriched in lymphocytes, and it may play a role in regulating lymphocyte homeostasis. This receptor has been shown to stimulate NF-kappa B activity and regulate cell apoptosis. The signal transduction of this receptor is mediated by various death domain containing adaptor proteins. Knockout studies in mice suggested the role of DR3/TNFRSF25 in the removal of self-reactive T cells in the thymus. Multiple alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported, most of which are potentially secreted molecules. The alternative splicing of this gene in B and T cells encounters a programmed change upon T-cell activation, which predomitly produces full-length, membrane bound isoforms, and is thought to be involved in controlling lymphocyte proliferation induced by T-cell activation.Specifications
DR3 | |
Human | |
IgG1, kappa | |
Monoclonal | |
Liquid | |
Antibody | |
4° C, store in dark, DO NOT FREEZE! |
JD3 | |
PE | |
Flow Cytometry | |
Primary | |
Monoclonal | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
RUO |
For Research Use Only.