missing translation for 'onlineSavingsMsg'
Learn More

DR3 Mouse anti-Human, PE, Clone: JD3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-6603-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15587066

  • £79.25 / 25 tests

Estimated Shipment
30-12-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

DR3/TNFRSF25 is a member of the TNF-receptor superfamily. This receptor is expressed preferentially in the tissues enriched in lymphocytes, and it may play a role in regulating lymphocyte homeostasis. This receptor has been shown to stimulate NF-kappa B activity and regulate cell apoptosis. The signal transduction of this receptor is mediated by various death domain containing adaptor proteins. Knockout studies in mice suggested the role of DR3/TNFRSF25 in the removal of self-reactive T cells in the thymus. Multiple alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported, most of which are potentially secreted molecules. The alternative splicing of this gene in B and T cells encounters a programmed change upon T-cell activation, which predomitly produces full-length, membrane bound isoforms, and is thought to be involved in controlling lymphocyte proliferation induced by T-cell activation.
Specifications

Specifications

DR3
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q93038
APO3; apoptosis inducing receptor; apoptosis-inducing receptor AIR; apoptosis-mediating receptor DR3; apoptosis-mediating receptor TRAMP; DDR3; death domain receptor 3 soluble form; death receptor beta; DR3; lymphocyte-associated receptor of death; protein WSL-1; TNFRSF12; tumor necrosis factor receptor superfamily member 25; tumor necrosis factor receptor superfamily, member 12; tumor necrosis factor receptor superfamily, member 12 (translocating chain-association membrane protein); tumor necrosis factor receptor superfamily, member 25; WSL; WSL1
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
JD3
PE
TNFRSF25
Liquid
TNFRSF25
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
8718
Videos
SDS
Documents

For Research Use Only.