Learn More
Dräger™ Short-Term Detector Tubes - De Tube N-hexane
Brand: Dräger™ 6728391
Code : 50
Additional Details : Weight : 0.07380kg
Description
1Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
2Safety Equipment Institute (SEI) certified.
Specifications
n-Hexane 100/a | |
100 to 3000ppm | |
10 | |
59°F to 86°F |
5 to 12mg/L | |
6 | |
3 min. | |
15°C to 30°C |