missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes: Carbon Dioxide

Detector tubes

Brand:  Dräger™ CH25101

Code : 50

Additional Details : CAS Number : 7664-93-9 Weight : 0.30000kg

 View more versions of this product

Product Code. 10615631

  • £61.35 / Pack of 10


Estimated Shipment
19-02-2018
Add to basket

Specifications

Specifications

Carbon dioxide
1 to 20%
1
0° to 40°C
40mg/L
30 sec.
10
Carbon dioxide 1%/a
Videos
Documentation