Learn More
Dräger™ Short-Term Detector Tubes: Toluene
Detector tubes
Brand: Dräger™ 8101731
Code : 50
Additional Details : CAS Number : 7664-93-9 Weight : 0.07380kg
Description
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Toluene | |
100 to 1800 | |
10 | |
32°F to 104°F | |
Toluene 100/a |
Less than30mg/L | |
10 | |
1.5 min. | |
0°C to 40°C |