missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ CMS Chips: Vinyl Chloride

Suitable for 10 measurements of one gas

Brand:  Dräger™ 6406170

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12344649

  • £91.40 / Each


Estimated Shipment
29-06-2018
Add to basket

Description

Description

Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications

Specifications

700 to 1100hpa
0.3 to 10
Vinyl Chloride CMS™ chip
1 to 30mg/L
30 to 420 sec.
0° to 40°C
Videos
Documentation