Learn More
DPY19L3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552545
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DTVELMNWIN SNTPRKAVFA GSMQLLAGVK LCTGRTLTNH PHYEDSSLRE RTRAVYQIYA KRAPEEVHAL LRSFGTDYVI LEDSICYERR HRRGCRLRDL LDIANGHMMD GPGENDPDLK PADHPRFCEE IKRNLP Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
DPY19L3 Polyclonal antibody specifically detects DPY19L3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
DPY19L3 | |
Polyclonal | |
Unconjugated | |
DPY19L3 | |
dpy-19 like 3 (C. elegans); DPY19L3; dpy-19-like 3; dpy-19-like protein 3; Probable C-mannosyltransferase DPY19L3; protein dpy-19 homolog 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
147991 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZPD9 | |
DPY19L3 | |
Recombinant protein corresponding to Human DPY19L3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only