missing translation for 'onlineSavingsMsg'
Learn More

DPPI 1c hydrochloride, Tocris Bioscience™

Dipeptidyl peptidase IV (DPP-IV) inhibitor

Brand:  Tocris (Bio-Techne) 278310

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13440594

  • £211.20 / 10mg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Inhibitor of dipeptidyl peptidase IV (DPP-IV) (IC50 = 104 nM) that displays selectivity over enzymes with DPP-like activity (IC50 > 30 μM). Increases plasma GLP-1 levels and improves glucose tolerance in diabetic mice following oral glucose challenge.
Specifications

Chemical Identifiers

866396-70-5
312.798
1-1-hydroxymethyl cyclopentyl amino acetyl-2,5-cis-pyrrolidinedicarbonitrile hydrochloride, 2s,5r-1-1-hydroxymethyl cyclopentyl glycyl pyrrolidine-2,5-dicarbonitrile hydrochloride
(2S,5R)-1-[2-[[1-(hydroxymethyl)cyclopentyl]amino]acetyl]pyrrolidine-2,5-dicarbonitrile;hydrochloride
C14H21ClN4O2
RHCWAHDCHQTNBR-IWKKHLOMSA-N
11381415
C1CCC(C1)(CO)NCC(=O)N2C(CCC2C#N)C#N.Cl

Specifications

DPPI 1c hydrochloride
866396-70-5
1-1-hydroxymethyl cyclopentyl amino acetyl-2,5-cis-pyrrolidinedicarbonitrile hydrochloride, 2s,5r-1-1-hydroxymethyl cyclopentyl glycyl pyrrolidine-2,5-dicarbonitrile hydrochloride
C1CCC(C1)(CO)NCC(=O)N2C(CCC2C#N)C#N.Cl
312.798
312.8
10mg
C14H21ClN4O2
RHCWAHDCHQTNBR-IWKKHLOMSA-N
(2S,5R)-1-[2-[[1-(hydroxymethyl)cyclopentyl]amino]acetyl]pyrrolidine-2,5-dicarbonitrile;hydrochloride
11381415
>98%
Videos