Learn More
DPPE fumarate, Tocris Bioscience™
Inhibitor of histamine binding at the intracellular binding site
Brand:  Tocris (Bio-Techne) 074350
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13427943
  • £279.30 / 50mg
Estimated Shipment date
from Supplier 01-04-2020
Description
Description
Binds with high affinity to the antiestrogen binding site but not to the estrogen receptor. Induces of sterol accumulation in MCF7 cells. An inhibitor of histamine binding at the intracellular histamine (Hlc) site.
Specifications
Chemical Identifiers
1185241-83-1
399.487
dppe fumarate, n,n-diethyl-2-4-phenylmethyl phenoxy ethanamine fumarate
2-(4-benzylphenoxy)-N,N-diethylethanamine;(E)-but-2-enedioic acid
C23H29NO5
ZOOMDYHTGZSGDD-WLHGVMLRSA-N
45073473
CCN(CC)CCOC1=CC=C(C=C1)CC2=CC=CC=C2.C(=CC(=O)O)C(=O)O
Specifications
50mg
1185241-83-1
dppe fumarate, n,n-diethyl-2-4-phenylmethyl phenoxy ethanamine fumarate
CCN(CC)CCOC1=CC=C(C=C1)CC2=CC=CC=C2.C(=CC(=O)O)C(=O)O
399.487
399.49
DPPE fumarate
C23H29NO5
ZOOMDYHTGZSGDD-WLHGVMLRSA-N
2-(4-benzylphenoxy)-N,N-diethylethanamine;(E)-but-2-enedioic acid
45073473
>98%