Learn More
DPN, Tocris Bioscience™
Highly potent ERβ agonist
Brand:  Tocris (Bio-Techne) 1494/10
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13439363
  • £132.30 / 10mg
Estimated Shipment
09-07-2019
Description
Description
Highly potent estrogen ERβ receptor agonist with a 70-fold selectivity over ERα (EC50 values are 0.85 and 66 nM respectively). Relaxes mesenteric arteries in vitro. Shown to regulate expression of GluR1, GluR2 and GluR3 in rat hippocampus.
Specifications
Chemical Identifiers
1428-67-7
239.274
diarylpropionitrile, 2,3-bis 4-hydroxyphenyl propionitrile, 2,3-bis 4-hydroxyphenyl propanenitrile, 2,3-bis 4-hydroxyphenyl-propionitrile, 2,3-bis p-hydroxyphenyl propionitrile, dpn compound, benzenepropanenitrile,4-hydroxy-a-4-hydroxyphenyl, diarylpropionitril, 2,3-bis p-hydroxyphenyl-propionitrile, propionitrile, 2,3-bis p-hydroxyphenyl
CHEBI:63949
C1=CC(=CC=C1CC(C#N)C2=CC=C(C=C2)O)O
C15H13NO2
GHZHWDWADLAOIQ-UHFFFAOYSA-N
102614
2,3-bis(4-hydroxyphenyl)propanenitrile
Specifications
DPN
1428-67-7
diarylpropionitrile, 2,3-bis 4-hydroxyphenyl propionitrile, 2,3-bis 4-hydroxyphenyl propanenitrile, 2,3-bis 4-hydroxyphenyl-propionitrile, 2,3-bis p-hydroxyphenyl propionitrile, dpn compound, benzenepropanenitrile,4-hydroxy-a-4-hydroxyphenyl, diarylpropionitril, 2,3-bis p-hydroxyphenyl-propionitrile, propionitrile, 2,3-bis p-hydroxyphenyl
C1=CC(=CC=C1CC(C#N)C2=CC=C(C=C2)O)O
239.274
CHEBI:63949
>99%
10mg
C15H13NO2
GHZHWDWADLAOIQ-UHFFFAOYSA-N
2,3-bis(4-hydroxyphenyl)propanenitrile
102614
239.27
Videos