Learn More
DPN, Tocris Bioscience™
Highly potent ERβ agonist
Brand:  Tocris (Bio-Techne) 149450
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13449363
  • £564.48 / 50mg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Highly potent estrogen ERβ receptor agonist with a 70-fold selectivity over ERα (EC50 values are 0.85 and 66 nM respectively). Relaxes mesenteric arteries in vitro. Shown to regulate expression of GluR1, GluR2 and GluR3 in rat hippocampus.
Specifications
Chemical Identifiers
1428-67-7
239.274
diarylpropionitrile, 2,3-bis 4-hydroxyphenyl propionitrile, 2,3-bis 4-hydroxyphenyl propanenitrile, 2,3-bis 4-hydroxyphenyl-propionitrile, 2,3-bis p-hydroxyphenyl propionitrile, dpn compound, benzenepropanenitrile,4-hydroxy-a-4-hydroxyphenyl, diarylpropionitril, 2,3-bis p-hydroxyphenyl-propionitrile, propionitrile, 2,3-bis p-hydroxyphenyl
CHEBI:63949
C1=CC(=CC=C1CC(C#N)C2=CC=C(C=C2)O)O
C15H13NO2
GHZHWDWADLAOIQ-UHFFFAOYSA-N
102614
2,3-bis(4-hydroxyphenyl)propanenitrile
Specifications
50mg
1428-67-7
diarylpropionitrile, 2,3-bis 4-hydroxyphenyl propionitrile, 2,3-bis 4-hydroxyphenyl propanenitrile, 2,3-bis 4-hydroxyphenyl-propionitrile, 2,3-bis p-hydroxyphenyl propionitrile, dpn compound, benzenepropanenitrile,4-hydroxy-a-4-hydroxyphenyl, diarylpropionitril, 2,3-bis p-hydroxyphenyl-propionitrile, propionitrile, 2,3-bis p-hydroxyphenyl
C1=CC(=CC=C1CC(C#N)C2=CC=C(C=C2)O)O
239.274
CHEBI:63949
>99%
DPN
C15H13NO2
GHZHWDWADLAOIQ-UHFFFAOYSA-N
2,3-bis(4-hydroxyphenyl)propanenitrile
102614
239.27