missing translation for 'onlineSavingsMsg'
Learn More

Doxorubicin hydrochloride, Tocris Bioscience™

Antitumor antibiotic agent. Inhibits DNA topoisomerase II

Brand:  Tocris (Bio-Techne) 225210

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13460874

  • £137.70 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Antitumor antibiotic agent that inhibits DNA topoisomerase II. DNA intercalator that inhibits nucleic acid synthesis and induces apoptosis. Reduces intracellular tau levels.
Specifications

Chemical Identifiers

25316-40-9
579.983
doxorubicin hydrochloride, adriamycin, doxorubicin hcl, adriacin, adriblastina, rubex, adriamycin hydrochloride, adriblastin, doxorubicin hydrochloride, adriamycin pfs
(7R,9R)-7-[(2S,4R,5R,6R)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
C27H30ClNO11
MWWSFMDVAYGXBV-BXPPNZEESA-N
129626538
CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)CO)O)N)O.Cl

Specifications

Doxorubicin hydrochloride
25316-40-9
doxorubicin hydrochloride, adriamycin, doxorubicin hcl, adriacin, adriblastina, rubex, adriamycin hydrochloride, adriblastin, doxorubicin hydrochloride, adriamycin pfs
CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)CO)O)N)O.Cl
579.983
579.99
10mg
C27H30ClNO11
MWWSFMDVAYGXBV-BXPPNZEESA-N
(7R,9R)-7-[(2S,4R,5R,6R)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
129626538
>98%
Videos