missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Door Marking Strips

Brand:  Brady™ 251948

Code : NEW

Additional Details : Weight : 0.05700kg

 View more versions of this product

Product Code. 11716655

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

1m x 60mm
Photoluminescent
Indicating doorways in the dark
1Each
Door Marking Strips
Videos