Learn More
Donkey anti-Rat IgG (H+L) Secondary Antibody, DyLight 405, Novus Biologicals™
Donkey Polyclonal Antibody
Brand:  Novus Biologicals NBP1-75651
153.00 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18508210
  • £195.00 / 1mg
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rat samples. It is validated for Flow Cytometry, Immunomicroscopy
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG (H+L)
Polyclonal
DyLight 405
10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
This Donkey anti-Rat IgG (H+L) Secondary Antibody [DyLight 405] was developed against purified rat IgG (H&L).
1 mg
Adaptive Immunity, Immunology
Reconstitute with sterilized deionized water. Centrifuge to remove any particulates.
Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage.
IgG
Flow Cytometry, Immunomicroscopy
LYOPH
Flow Cytometry 1:20 - 1:2000, Immunomicroscopy 1:20 - 1:2000
Donkey
Affinity purified
RUO
Secondary
Rat
Purified