Learn More
Donkey anti-Chicken IgY (H+L) Secondary Antibody, Biotin, Invitrogen™
Donkey Polyclonal Secondary Antibody
Brand: Invitrogen SA172003
126.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LD
Additional Details : Weight : 0.01000kg
Description
SA1-72003 detects Chicken IgY in chicken samples. SA1-72003 has been successfully used in ELISA and western blot procedures. The SA1-72003 immunogen is from purified egg-derived IgY.
Chicken IgY (H+L) Polyclonal antibody specifically detects Chicken IgY (H+L) in Chicken samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
Chicken IgY (H+L) | |
Polyclonal | |
Biotin | |
Donkey | |
Affinity chromatography | |
RUO | |
Chicken | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1.0 mg/mL | |
PBS with 50% glycerol and 0.1% chloroacetamide; pH 7.2 | |
Purified egg-derived IgY. | |
500 μg | |
Secondary | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG |