missing translation for 'onlineSavingsMsg'
Learn More

Donepezil hydrochloride, Tocris Bioscience™

Potent AChE inhibitor

Brand:  Tocris (Bio-Techne) 438550

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13434664

  • £286.65 / 50mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent acetylcholinesterase inhibitor (IC50 = 5.7 nM). Displays 1252-fold selectivity for AChE over BuChE (IC50 = 7138 nM for BuChE). Orally active.

Deuterated analog also available.

Specifications

Chemical Identifiers

120011-70-3
415.958
donepezil hydrochloride, donepezil hcl, aricept, bnag, donepezilhcl, aricept odt, 2-1-benzylpiperidin-4-yl methyl-5,6-dimethoxy-2,3-dihydro-1h-inden-1-one hydrochloride, e 2020 pharmaceutical, eranz
CHEBI:4696
COC1=C(C=C2C(=C1)CC(C2=O)CC3CCN(CC3)CC4=CC=CC=C4)OC.Cl
C24H30ClNO3
XWAIAVWHZJNZQQ-UHFFFAOYSA-N
5741
2-[(1-benzylpiperidin-4-yl)methyl]-5,6-dimethoxy-2,3-dihydroinden-1-one;hydrochloride

Specifications

50mg
120011-70-3
donepezil hydrochloride, donepezil hcl, aricept, bnag, donepezilhcl, aricept odt, 2-1-benzylpiperidin-4-yl methyl-5,6-dimethoxy-2,3-dihydro-1h-inden-1-one hydrochloride, e 2020 pharmaceutical, eranz
COC1=C(C=C2C(=C1)CC(C2=O)CC3CCN(CC3)CC4=CC=CC=C4)OC.Cl
415.958
CHEBI:4696
>99%
Donepezil hydrochloride
C24H30ClNO3
XWAIAVWHZJNZQQ-UHFFFAOYSA-N
2-[(1-benzylpiperidin-4-yl)methyl]-5,6-dimethoxy-2,3-dihydroinden-1-one;hydrochloride
5741
415.95
Videos
Documents