missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donepezil hydrochloride, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 211.0°C to 212.0°C |
Infrared Spectrum | Authentic |
Quantity | 1 g |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in hot methanol, insoluble in ethyl acetate and hexane |
Formula Weight | 415.85 |
Percent Purity | ≥97.5% (HPLC) |
Chemical Name or Material | Donepezil hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'