Learn More
Domoic acid, Tocris Bioscience™
Brand:  Tocris (Bio-Techne) 0269/1
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 1mg
 View more versions of this product
Product Code. 13487513
  • £157.08 / 1mg
Estimated Shipment
04-10-2018
Add to basket
Description
Description
Kainate receptor agonist. More potent and possibly more selective than kainate at kainate receptors, as demonstrated in electrophysiological studies.
Specifications
Chemical Identifiers
14277-97-5
311.334
(2S,3S,4R,5'R)-2-Carboxy-4-(5'-carboxy-1'-methyl-1Z,3E-hexadienyl)-3-pyrrolidineacetic acid
CHEBI:34727
C15H21NO6
VZFRNCSOCOPNDB-AOKDLOFSSA-N
5282253
Specifications
Domoic acid
14277-97-5
C15H21NO6
VZFRNCSOCOPNDB-AOKDLOFSSA-N
(2S,3S,4S)-4-[(2Z,4E,6R)-6-carboxyhepta-2,4-dien-2-yl]-3-(carboxymethyl)pyrrolidine-2-carboxylic acid
5282253
311.33
1mg
>95%
(2S,3S,4R,5'R)-2-Carboxy-4-(5'-carboxy-1'-methyl-1Z,3E-hexadienyl)-3-pyrrolidineacetic acid
CC(C=CC=C(C)C1CNC(C1CC(=O)O)C(=O)O)C(=O)O
311.334
CHEBI:34727
Videos