Thermo Scientific™ Domed PCR Cap Strips

Optimize PCR applications with these cap strips for use with Thermo Scientific 96-well PCR plates and strip tubes.

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: AB-0786

UNSPSC: 12352200

Code: 04

Additional Details:
Additional Details: Weight: 1.00000kg



Product Code. 11574794

Quantity Price
1 £ 51.25 / Pack of 300
EU Stock 14
Add to basket


Description and Specification

Specification

No. per Pack 12
For Use With 96-well PCR plates as well as standard and low profile PCR Tube Strips
Format Strips of 8 Flat Caps
Quantity 25 packs of 12
Color Natural
Well Count 96
Material Virgin Medical Grade Polypropylene
Applications Facilitates sample labeling
Certifications/Compliance Certified free of human DNA, RNase, DNase, and endotoxins.
No. per Case 300

Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.

Highlights:
  • Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
  • Large end tabs for strip removal (12 strips only)
Certifications:
Certified free from human DNA, RNases, and DNases.

Documentation

Technical documents