missing translation for 'onlineSavingsMsg'
Learn More

Dodecyltrimethylammonium chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-3778500
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
112-00-5
C15H34ClN
263.88
MFCD00041974
DDXLVDQZPFLQMZ-UHFFFAOYSA-M
dodecyltrimethylammonium chloride, alicop, cation bb, aliquat 4, n,n,n-trimethyldodecan-1-aminium chloride, laurtrimonium chloride, dehyquart lt, cation fb, nissan cation bb, lauryltrimethylammonium chloride
8152
dodecyl(trimethyl)azanium;chloride
CCCCCCCCCCCC[N+](C)(C)C.[Cl-]
This item is not returnable. View return policy
112-00-5
C15H34ClN
263.88
MFCD00041974
DDXLVDQZPFLQMZ-UHFFFAOYSA-M
dodecyltrimethylammonium chloride, alicop, cation bb, aliquat 4, n,n,n-trimethyldodecan-1-aminium chloride, laurtrimonium chloride, dehyquart lt, cation fb, nissan cation bb, lauryltrimethylammonium chloride
8152
dodecyl(trimethyl)azanium;chloride
CCCCCCCCCCCC[N+](C)(C)C.[Cl-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY
Melting Point 237.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3(CH2)11N(CH3)3Cl
Quantity 25 g
Solubility Information (10% in water) Clear
Formula Weight 263.88
Percent Purity 98%
Physical Form Chunks and/or Powder
Chemical Name or Material Dodecyltrimethylammonium chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dodecyltrimethylammonium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'