missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Dodecacarbonyltetrairidium, 98%

Brand:  Alfa Aesar™ 010505.03

Code : NEW

Additional Details : CAS Number : 11065-24-0 Weight : 0.00100kg Transport : UN number : 3466 Chem class : 1 Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 11395998

  • £299.90 / 1g


Estimated Shipment date
from Supplier 26-02-2018
Add to basket

Specifications

Chemical Identifiers

Dodecacarbonyltetrairidium
1105.01
UN3466
11065-24-0
MFCD00011064
Tetrairidium dodecacarbonyl; Iridium carbonyl

Specifications

Dodecacarbonyltetrairidium
1g
Powder
1105.01
UN3466
195°C (dec.)
Air Sensitive
11065-24-0
98%
MFCD00011064
Tetrairidium dodecacarbonyl; Iridium carbonyl
Videos
SDS