Learn More
Docetaxel, Tocris Bioscience™
Microtubule stabilizer
Brand:  Tocris (Bio-Techne) 405610
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13434054
  • £144.06 / 10mg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
Binds and stabilizes microtubules; leads to cell cycle arrest and apoptosis. Displays significant antitumor activity in ovarian and gastric tumors. Prevents microtubule depolymerization and disassembly in the absence of GTP. Exhibits cytotoxic activity.
Specifications
Chemical Identifiers
114977-28-5
807.89
docetaxel, taxotere, docetaxel anhydrous, docetaxol, emdoc, docetaxel, trihydrate, docetaxel inn, taxotere tn, unii-699121phca, txl
CHEBI:4672
C43H53NO14
ZDZOTLJHXYCWBA-VCVYQWHSSA-N
148124
CC1=C2C(C(=O)C3(C(CC4C(C3C(C(C2(C)C)(CC1OC(=O)C(C(C5=CC=CC=C5)NC(=O)OC(C)(C)C)O)O)OC(=O)C6=CC=CC=C6)(CO4)OC(=O)C)O)C)O
Specifications
10mg
114977-28-5
docetaxel, taxotere, docetaxel anhydrous, docetaxol, emdoc, docetaxel, trihydrate, docetaxel inn, taxotere tn, unii-699121phca, txl
CC1=C2C(C(=O)C3(C(CC4C(C3C(C(C2(C)C)(CC1OC(=O)C(C(C5=CC=CC=C5)NC(=O)OC(C)(C)C)O)O)OC(=O)C6=CC=CC=C6)(CO4)OC(=O)C)O)C)O
148124
807.88
Docetaxel
C43H53NO14
ZDZOTLJHXYCWBA-VCVYQWHSSA-N
807.89
CHEBI:4672
>99%