missing translation for 'onlineSavingsMsg'
Learn More

TCR DO11.10 Mouse anti-Mouse, PE-Cyanine7, Clone: KJ1-26 (KJ126, KJI-26, KJI26), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 25-5808-80

Code : NEW

Additional Details : Weight : 0.09500kg

Product Code. 15578566

  • £85.50 / 25µg

Estimated Shipment
30-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. TCR DO11 (DO11.10) is a receptor found on lymphocytes of DO11.10 transgenic mice and T-cell receptors of BALB/c-derived DO11.10 and DO11.10.24 T cell hybridomas. The DO11.10 TCR recognizes a C-terminal ovalbumin peptide (OVA 323-339) in the context of I-Ad major histocompatibility molecules. The use of antibodies against DO11.10 have proven to be instrumental in describing many of the intricate details of T cell immunity, including T cell activation, development, and function.
Specifications

Specifications

TCR DO11.10
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
D011.10; T-cell receptor DO11.10
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
Mouse
Flow Cytometry
KJ1-26 (KJ126, KJI-26, KJI26)
PE-Cyanine7
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Videos
Documents

For Research Use Only.