Learn More
Thermo Scientific Chemicals DNAzol™ Reagent
DNAzol|r Reagent, is a complete and ready-to-use electrophoretic reagent to isolate genomic DNA from solid and liquid samples of animal, plant, yeast, and bacterial origin.
Brand: Thermo Scientific Chemicals J60637.AE
Code : NEW
Additional Details : Weight : 0.00010kg
Description
DNAzol™ Reagent is used as an electrophoretic reagent for the isolation of genomic DNA from solid and liquid samples of animal and plant origin
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
- Cholesterol (cholesterin, cholesteryl alcohol) is a lipid sterol found in vertebrate cell membranes
- Cholesterol is a cholestanoid, a class of molecule that contains the C-27 cholestane motif
- A precursor of many steroids and sex hormones in animals including cortisol, aldosterone, testosterone, progesterone, and estrogen
- It plays a role in neurotransmission and is crucial for organ systems, such as immune, reproductive and endocrine systems
Applications
- Suitable for in vitro and in vivo applications
- Stabilized in powder form for laboratory use only
Specifications
Blue-green | |
DNA Extraction Reagent | |
Liquid |
Complete and ready to use reagent for the isolation of genomic DNA from solid and liquid samples of animal and plant origin | |
100 mL |
RUO – Research Use Only