missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DNase I, RNase-free

An endonuclease that nonspecifically cleaves single- and double-stranded DNA to release di-, tri-, and oligonucleotides with 5'-phosphate and 3'-OH groups.

£48.48 - £49.21

Specifications

Inhibitors metal chelators, transition metals (e.g., Zn) in millimolar concentrations, SDS (even at concentrations less than 0.1%), reducing agents (DTT and beta-mercaptoethanol), ionic strength above 50-100 mM
Molecular Weight 29 kDa monomer
Source E.coli cells with a cloned gene encoding bovine DNase
Quantity 1000U
View More Specs

Products 2
Product Code Brand Quantity Concentration Price Quantity & Availability  
Product Code Brand Quantity Concentration Price Quantity & Availability  
10649890 Thermo Scientific™
EN0521
1000U 1U/μL £48.48
1000 units View alternative products
Save Up to 
»
EU Stock 189 Please to purchase this item. Need a web account? Register with us today!
10272490 Thermo Scientific™
EN0523
1000U 50U/μL £49.21
1000 units View alternative products
Save Up to 
»
EU Stock 106 Please to purchase this item. Need a web account? Register with us today!
Description

Description

Highlights

  • Recombinant enzyme
  • Purified from non-animal host with a lower level of intrinsic RNases
Specifications

Specifications

metal chelators, transition metals (e.g., Zn) in millimolar concentrations, SDS (even at concentrations less than 0.1%), reducing agents (DTT and beta-mercaptoethanol), ionic strength above 50-100 mM
29 kDa monomer
E.coli cells with a cloned gene encoding bovine DNase
1000U