missing translation for 'onlineSavingsMsg'
Learn More

DNase I from Bovine Pancreas, Thermo Scientific™

Derived from Bovine pancreas.

Brand:  Thermo Scientific J14340.F+

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15845278

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Synonym: E.C.3.1.21.1
  • MDL: MFCD00130917
Specifications

Specifications

Cell Lysis Enzyme
Bovine Pancreas
Lyophilized
20 mg
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only