missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DNAPac™ PA100 Oligonucleotide HPLC Columns

Features strong anion-exchange resin for high-resolution analysis and purification of synthetic and modified oligonucleotides.

Brand:  Thermo Scientific™ 088764

Additional Details : Weight : 0.55000kg

 View more versions of this product

Product Code. 15540224

  • £779.00 / Each


Estimated Shipment: 03-11-2021
Add to basket

Description

Description

Feature strong anion-exchange resin for high-resolution analysis and purification of synthetic and modified oligonucleotides with Thermo Scientific™ Dionex™ DNAPac™ PA100 Oligonucleotide Columns.

Achieve n, n-1 resolution for oligonucleotides

  • Resolve oligonucleotides with secondary structures
Specifications

Specifications

Preparative Column, Standard
1.5 mL/min
4000 psi (275 bar)
2 to 12.5
90°C
DNAPac
Ion Exchange
Nucleic Acid/Anion Exchange
9 mm
50 mm
13 μm
Nonporous
Quaternary Ammonium
Spherical, Nonporous Polymer
Ion Exchange
Videos
SDS
Documents
Certificates

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.