Learn More
DNAJC7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554667
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ATLMMLGRFR EALGDAQQSV RLDDSFVRGH LREGKCHLSL GNAMAACRSF QRALELDHKN AQAQQEFKNA NAVME Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
DNAJC7 Polyclonal antibody specifically detects DNAJC7 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
DNAJC7 | |
Polyclonal | |
Unconjugated | |
DNAJC7 | |
2010003F24Rik; 2010004G07Rik; CCRP; Cytoplasmic CAR retention protein; cytoplasmic constitutive active/androstane receptor retention protein; DJ11; DJC7; DnaJ (Hsp40) homolog, subfamily C, member 7; DnaJ heat shock protein family (Hsp40) member C7; dnaJ homolog subfamily C member 7; DNAJC7; MDj11; mTpr2; tetratricopeptide repeat domain 2; tetratricopeptide repeat protein 2; TPR repeat protein 2; TPR2; TTC2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
303536, 56354, 7266 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99615, Q9QYI3 | |
DNAJC7 | |
Recombinant protein corresponding to Human DNAJC7 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only