missing translation for 'onlineSavingsMsg'
Learn More

DMPQ dihydrochloride, Tocris Bioscience™

Potent, selective inhibitor of PDGFRβ

Brand:  Tocris (Bio-Techne) 122250

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13438643

  • £393.92 / 50mg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A potent and selective inhibitor of human vascular β-type platelet derived growth factor receptor tyrosine kinase (PDGFRβ) (IC50 = 80 nM). Displays > 100-fold selectivity over EGFR tyrosine kinase, erbB2, p56, protein kinase A and protein kinase C.
Specifications

Chemical Identifiers

1123491-15-5
339.216
dmpq dihydrochloride, 5,7-dimethoxy-3-4-pyridinyl quinoline dihydrochloride, 5,7-dimethoxy-3-pyridin-4-yl quinoline dihydrochloride, dmpq hydrochloride, dmpq dihydrochloride hplc, 5,7-dimethoxy-3-4-pyridinyl quinolinedihydrochloride
5,7-dimethoxy-3-pyridin-4-ylquinoline;dihydrochloride
C16H16Cl2N2O2
YBBAOKYVJCNJIV-UHFFFAOYSA-N
45073427
COC1=CC2=NC=C(C=C2C(=C1)OC)C3=CC=NC=C3.Cl.Cl

Specifications

DMPQ dihydrochloride
1123491-15-5
dmpq dihydrochloride, 5,7-dimethoxy-3-4-pyridinyl quinoline dihydrochloride, 5,7-dimethoxy-3-pyridin-4-yl quinoline dihydrochloride, dmpq hydrochloride, dmpq dihydrochloride hplc, 5,7-dimethoxy-3-4-pyridinyl quinolinedihydrochloride
COC1=CC2=NC=C(C=C2C(=C1)OC)C3=CC=NC=C3.Cl.Cl
339.216
339.22
50mg
C16H16Cl2N2O2
YBBAOKYVJCNJIV-UHFFFAOYSA-N
5,7-dimethoxy-3-pyridin-4-ylquinoline;dihydrochloride
45073427
>99%
Videos